missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C5a Antibody (C17/5), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23498JF646
This item is not returnable.
View return policy
Description
Complement Component C5a Monoclonal antibody specifically detects Complement Component C5a in Human, Baboon samples. It is validated for Western Blot, ELISA
Specifications
| Complement Component C5a | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Baboon, Rat | |
| Purified |
| Western Blot, ELISA | |
| C17/5 | |
| Western Blot, ELISA | |
| C5, C5A, HE | |
| Purified human C5a | |
| 0.1 mL | |
| Immunology, Innate Immunity, Stem Cell Markers | |
| 727 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction