Learn More
Novus Biologicals™ Complement Component C5aR1 Antibody (G2 patent anti-C5aR) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28649-1mg
 View more versions of this product
Product Code. 30047172
  • € 920.00 / 1mg
Description
Complement Component C5aR1 Monoclonal antibody specifically detects Complement Component C5aR1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Complement Component C5aR1
Monoclonal
Unconjugated
P21730
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
G2 patent anti-C5aR
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
C5A, C5a anaphylatoxin chemotactic receptor, C5aR, C5a-R, C5ARC5a anaphylatoxin receptor, C5R1C5a ligand, CD88, CD88 antigen, complement component 5 receptor 1, complement component 5 receptor 1 (C5a ligand), complement component 5a receptor 1
Complement C5aR1
1 mg
GPCR, Immunology, Innate Immunity
728