missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C9 Antibody (22), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-21612AF750
This item is not returnable.
View return policy
Description
Complement Component C9 Monoclonal specifically detects Complement Component C9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Complement Component C9 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| C9 | |
| Complement factor C9 adsorbed onto aluminum hydroxide gel. | |
| Primary | |
| Human | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 22 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| complement component 9, complement component C9 | |
| Mouse | |
| 0.1 mL | |
| 735 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction