missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Complement Factor D/Adipsin Antibody (27) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26908AF750
This item is not returnable.
View return policy
Description
Complement Factor D/Adipsin Monoclonal antibody specifically detects Complement Factor D/Adipsin in Human samples. It is validated for ELISA
Specifications
| Complement Factor D/Adipsin | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Adipsin, ADNcomplement factor D, C3 convertase activator, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement Factor D/Adipsin (Uniprot#: P00746; Ile26-Ala253). | |
| 0.1 mL | |
| Immunology | |
| 1675 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 27 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction