missing translation for 'onlineSavingsMsg'
Learn More

Complement factor D-IN-1, MedChemExpress

Product Code. 16416437
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16416437

Brand: Medchem Express HY102034

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Complement factor D-IN-1 is a potent and selective small-molecule reversible factor d inhibitor, with IC50s of 0.006 and 0.05 μM in FD Thioesterolytic Fluorescent Assay and a MAC Deposition Assay, respectively.

Complement factor D-IN-1 is a potent and selective small-molecule reversible factor d inhibitor, with IC50s of 0.006 and 0.05 μM in FD Thioesterolytic Fluorescent Assay and a MAC Deposition Assay, respectively.

Specifications

Chemical Name or Material Complement factor D-IN-1
CAS 1386455-76-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H19BrN6O3
Quantity 5 mg
Solubility Information DMSO : 250 mg/mL (517.26 mM; Need ultrasonic)
SMILES O=C(N)C1=NN(CC(N2[C@H](C(NC3=CC=CC(Br)=N3)=O)C[C@@H]4[C@H]2C4)=O)C5=C1C=CC=C5
Molecular Weight (g/mol) 483.32
Formula Weight 483.32
Percent Purity 98.65%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.