missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Complement Factor H Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524425
This item is not returnable.
View return policy
Description
Complement Factor H Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Factor H is a serum protein acting as an inhibitor in the complement activation system. I activates the C3/C5 convertase by dissociating its subunits and also acts as a cofactor for the enzyme activity of Factor I.
Specifications
| Complement Factor H | |
| Polyclonal | |
| Unconjugated | |
| CFH | |
| adrenomedullin binding protein; age-related maculopathy susceptibility 1; AHUS1; AMBP1; ARMD4; ARMS1; beta-1H; beta-1-H-globulin; Cfh; CFHL3; complement component factor H; complement factor H; complement factor H related protein 3A4/5G4; complement factor H, isoform b; complement inhibitory factor H; factor H; factor H-like 1; FH; FHL1; FHL1 antibody; H factor 1; H factor 1 (complement); H factor 2 (complement); HF; HF1; HF2; HGNC:4883; HUS; MGC88246; Mud-1; NOM; platelet complement factor H; protein beta-1-H; RP1-177P10.1; Sas1; Sas-1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 3075 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P08603 | |
| CFH | |
| KLH conjugated synthetic peptide between 751-780 amino acids of human CFH. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction