missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Complement Factor H-related 1/CFHR1/CFHL1 Recombinant Protein Antigen

Product Code. 18176694 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
This item is not returnable. View return policy

Product Code. 18176694

Brand: Novus Biologicals™ NBP214475PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CFHR1. The Complement Factor H-related 1/CFHR1/CFHL1 Recombinant Protein Antigen is derived from E. coli. The Complement Factor H-related 1/CFHR1/CFHL1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP2-14475. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 3078
Species Human
Purification Method Chromatography
Purity >80%
Concentration 0.5mg/mL
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Symbol CFHR1
Label Type Unlabeled
Molecular Weight (g/mol) 22kDa
Product Type Complement Factor H-related 1/CFHR1/CFHL1
Quantity 0.1 mL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-14475. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogen YKPFSQVPTGEVFYYSCEYNFVSPSKSFWTRITCTEEGWS
Mostrar más Mostrar menos

For Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado