Learn More
Complement Factor H-related 2/CFHR2/CFHL2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-00224MFV450
Product Code. 18578779
  • € 428.00 / 0.10mL
Description
Complement Factor H-related 2/CFHR2/CFHL2 Polyclonal antibody specifically detects Complement Factor H-related 2/CFHR2/CFHL2 in Human samples. It is validated for ELISA