Learn More
Complement Factor H-related 4/CFHR4 Antibody (640212), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB5980U
Product Code. 30066261
  • € 607.00 / 0.10mL
Description
Complement Factor H-related 4/CFHR4 Monoclonal antibody specifically detects Complement Factor H-related 4/CFHR4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Complement Factor H-related 4/CFHR4
Monoclonal
Alexa Fluor 350
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
10877
Store at 4C in the dark.
IgG1
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
640212
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
complement factor H-related protein 4, CFHL4, complement factor H-related 4, FHR4, FHR-4
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Complement Factor H-related 4/CFHR4, Glu20-Glu331, Accession # Q92496
0.1 mL