Learn More
Goat IgG Antibody (GT175), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50413H
Product Code. 18616838
  • € 554.00 / 0.10mL
Description
Goat IgG Monoclonal antibody specifically detects Goat IgG in Goat samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Goat IgG
Monoclonal
HRP
Immunoglobulin G
Goat immunoglobulins
0.1 mL
Primary
Store at 4°C in the dark.
IgG1
Immunohistochemistry
GT175
PBS
Mouse
Protein A purified
RUO
Goat