missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Complement Factor I Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575509
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Factor I, a serine protease, is a regulatory component of both the alternative and the classical complement pathway. Factor I is synthesized as a single chain precursor of 88 kDa, processed to yield the active form, composed of two disulfide-linked chains, the a-chain (50 kDa) and the b-chain (38 kDa). Plasma concentration is 35 μg/mL.
Specifications
| Complement Factor I | |
| Polyclonal | |
| Unconjugated | |
| CFI | |
| AHUS3; ARMD13; C3B/C4B inactivator; C3BINA; C3b-INA; C3b-inactivator; Cfi; complement component factor i; complement component I; complement control protein factor I; complement factor I; Complement factor I heavy chain; Complement factor I light chain; factor i; FI; IF; KAF; Konglutinogen-activating factor; light chain of factor I | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12630, 3426, 79126 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P05156, Q61129, Q9WUW3 | |
| CFI | |
| Synthetic peptide corresponding to amino acids 492-537 of Human Complement Factor I. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction