missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor MASP3 Antibody (MM0197-4F19), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12204F
This item is not returnable.
View return policy
Description
Complement Factor MASP3 Monoclonal antibody specifically detects Complement Factor MASP3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Complement Factor MASP3 | |
| Monoclonal | |
| FITC | |
| Complement-activating component of Ra-reactive factor, CRARF, Mannan-binding lectin serine protease 1 isoform 2, Mannose-binding protein-associated serine protease 3, MASP3, RaRF | |
| Human recombinant Complement Factor MASP3 (Catalytic Domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0197-4F19 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 5648 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction