Learn More
Agilent BioTek Complete Dispense and Waste Systems
Product Code. 16522192 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
50 TS Microplate Washer, Agilent BioTek Microplate Washer and Reader Instruments
50 TS with Automated Buffer Switching Capabilities, Agilent BioTek Microplate Washer and Reader Instruments
Unit Size:
Each
Product Code. 16522192
Brand: Agilent BioTek 4070078
Complete Dispense and Waste Systems include waste and dispense accessories, including racks and tubing, and are for use with various models of microplate washers
4070077 includes: 3 Supply Bottles with Filter, 4 L; 1 Waste Bottle, 4 L; 2 Racks, and Tubing, 4070078 includes: 2 Dispense Bottles with Filter, 4L; 1 Waste Bottle, 4L ; 2 Racks, and Tubing
TRUSTED_SUSTAINABILITY
Specifications
Type Waste System
For Use With (Equipment) 50 TS Microplate Washer, Agilent BioTek Microplate Washer and Reader Instruments