missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMT Rabbit anti-Human, Rat, Clone: 9Y2A3, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 190.00 - € 496.00
Specifications
| Antigen | COMT |
|---|---|
| Clone | 9Y2A3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18304752
|
Bio-Techne
NBP3-16413-20UL |
20 μg |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18302932
|
Bio-Techne
NBP3-16413-100UL |
100 μg |
€ 496.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COMT Monoclonal antibody specifically detects COMT in Human, Rat samples. It is validated for Western BlotSpecifications
| COMT | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1312 | |
| IgG | |
| Affinity purified |
| 9Y2A3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| catechol O-methyltransferase, catechol-O-methyltransferase, EC 2.1.1.6 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human COMT (P21964). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title