Learn More
Motic™ Polarizer, AE2000MET Series
Improve image contrast with a simple polarization set containing a polarizer and analyzer. Motic™ Polarizer AE2000MET Series can be rotated 360° to accommodate observation demands.
Brand:  Motic™ 1101000202111
Additional Details : Weight : 0.01000kg
Product Code. 15455332
  • € 45.00 / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket
Description
Description
  • Improve image quality obtained with birefringent materials when compared to other techniques such as darkfield and brightfield illumination, differential interference contrast, phase contrast, Hoffman modulation contrast, and fluorescence.
  • Useful for both quantitative and qualitative studies targeted at a wide range of anisotropic specimens.
  • Polarized light microscopy is capable of providing information on absorption color and optical path boundaries between minerals of differing refractive indices, in a manner similar to brightfield illumination, but the technique can also distinguish between isotropic and anisotropic substances.
  • Contrast-enhancing technique exploits the optical properties specific to anisotropy and reveals detailed information concerning the structure and composition of materials that are invaluable for identification and diagnostic purposes.
Specifications
Specifications
Polarizer
AE2000MET Series Microscopes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers