Learn More
1413 μS/cm Conductivity Standard Solution Sachets, Hanna Instruments™
Click to view available options
Quantity:
500 mL
25 x 20 mL
Packaging:
Bottles
Light Block Bottles
Sachets
Unit Size:
460mL
Each
Description
- A premium quality 1,413 μS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution and supplied with a certificate of analysis.
- Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
- Air tight bottle with tamper-proof seal to ensure quality
- Premium quality 1,413 μS/cm conductivity solution standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material)
- Deionized water for analytical use in accordance with ISO 3696/BS3978
- Additional equipment used in preparation of the solutions include certified weight-checked balances, Class A glassware and certified thermometers
Specifications
Specifications
| Packaging | Bottles |
| Odor | Odorless |
| Physical Form | Liquid |
| Color | Colorless |
| pH | 7 |
| Solubility | Soluble |
| Quantity | 500 mL |