Learn More
Conduritol B Tetraacetate, TRC
Specifications
Specifications
| Chemical Name or Material | Conduritol B Tetraacetate |
| CAS | 25348-63-4 |
| Molecular Formula | C14 H18 O8 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C14H18O8/c1-7(15)19-11-5-6-12(20-8(2)16)14(22-10(4)18)13(11)21-9(3)17/h5-6,11-14H,1-4H3/t11-,12+,13+,14- |
| SMILES | CC(=O)O[C@@H]1C=C[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C |
| IUPAC Name | [(1R,4S,5R,6S)-4,5,6-triacetyloxycyclohex-2-en-1-yl] acetate |
| Molecular Weight (g/mol) | 314.29 |
| Formula Weight | 314.1002 |