Learn More
Greiner Bio-One Conical Bottom Polypropylene Test Tubes with Screw Cap
GREENER_CHOICE
Product Code. p-7075352
Click to view available options
Capacity (Metric):
50 mL
Closure Color:
Blue
Product Type:
Centrifuge Tube with Support Skirt
Quantity:
300/Cs.
Unit Size:
Pack of 300
Product Code. 10529121
Brand: Greiner Bio-One 210270
Only null left
Add to basket
Add to basket
For centrifugation and applications that require chemical and thermo resistant vessels.
  • Natural colored tube
  • Blue graduations
  • White writing area
  • With or without support skirt
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Color Natural
Capacity (Metric) 50 mL
Material Polypropylene
For Use With Centrifugation and applications that require chemical and thermo resistant vessels.
No. per Case 300
Quantity 300/Cs.
Sterility Sterile
Length (Metric) 115 mm
Bottom Shape Conical
Writing Area White Marking Spot
Diameter (Metric) 30 mm
Graduated Yes
Product Type Centrifuge Tube with Support Skirt