missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Conivaptan Hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Conivaptan hydrochloride is a very high affinity vasopressin V1A and V2 antagonist (Ki values are 0.61 and 0.66 - 3.04 nM, respectively). Inhibits vasopressin-induced pressor responses in rats without altering blood pressure, and increases urine flow while decreasing urine osmolality. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N-[4-[(4,5-Dihydro-2-methylimidazo[4,5-d][benzazepin-6(1H)-yl)carbonyl]phenyl-[1,1'-biphenyl]-2-carboxamide hydrochloride |
| CAS | 168626-94-6 |
| Quantity | 50 mg |
| Molecular Formula | C32H26N4O2.HCl |
| Formula Weight | 535.04 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction