Learn More
BOLA Connection Bolts
Click to view available options
Includes:
Fingertight Nut, Single Ferrule
Hex-head Nut, Double Ferrule
Unit Size:
Each
Description
- Connection bolt and sealing cone made of PEEK, suitable for female thread UNF 10-32 G.
- Exchangeable sealing cone available separately, knurled nut reusable.
- Suitable for pressures up to max.
- 280 bar
Specifications
Specifications
| Diameter (English) Outer | 1.6 in. |
| Includes | Fingertight Nut, Single Ferrule |
| Autoclavable | Autoclavable |