Learn More
WTW™ Connection Cable for Depth Probes
Product Code. p-8001942
Click to view available options
Length (Metric):
10 m
15 m
3 m
6 m
Unit Size:
Each
Product Code. 15100173
Brand: WTW™ 903854
Only null left
Add to basket
Add to basket
For use with WTW™ MPP IDS depth probes with waterproof plug head.
  • Also useful for plug head sensors
  • Available in a range of lengths
  • Supplied individually
TRUSTED_SUSTAINABILITY
Specifications
Type Connection Cable
Length (Metric) 15 m
For Use With (Equipment) MPP IDS with waterproof head sensors