Learn More
SI Analytics™ Connection Cables
Product Code. p-7160702 Shop All SI Analytics Products
Click to view available options
Type:
B 1 N
K 5 S HT
Unit Size:
Each
Product Code. 10557291
Brand: SI Analytics™ 285124354
Only null left
Add to basket
Add to basket
SI Analytics™ Connection Cables are available in various combinations of electrode socket, instrument plug, and cable lengths.
  • Electrode plug for reference electrode
  • With extension cable
TRUSTED_SUSTAINABILITY
Specifications
Description Coaxial Electrode Plug
Type K 5 S HT
Includes Electrode socket and connector plug
Temperature (Metric) Operating 135°C
Length (Metric) Cable 5 m