Learn More
JULABO™ Connection Kit for Fluid-Bypass
Includes all necessary components to fix the Fluid-Bypass to the PRESTO™ model.
Brand: JULABO™ 8980781
Additional Details : Weight : 6.00000kg
Description
- Stainless steel tubing to connect Fluid-Bypass to PRESTO™
- Insulation, temperature resistant
- Digital control cable
Specifications
6 kg | |
Connection Kit for Fluid-Bypass | |
For Julabo™ Fluid-Bypass, Electronically Controlled | |
PRESTO™ A45, A45t, W50, W50t |