Learn More
Lenz Laborglasintrumente Connectors with Stopcock
Product Code. 10478402
Click to view available options
Standard Taper Joint No.:
Socket NS 14/23, Cone NS 14/23
Socket NS 29/32, Cone NS 29/32
Unit Size:
Each
Product Code. 10478402
Brand: Lenz Laborglasintrumente 05310044
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Connectors with NS stopcock
  • With solid glass plug
  • With NS socket and NS cone
  • Made of DURAN™ tubing
Specifications
Material Duran Glass
Standard Taper Joint No. Socket NS 29/32, Cone NS 29/32