Learn More
Lenz Laborglasintrumente Connectors with Stopcock
Click to view available options
Standard Taper Joint No.:
Socket NS 14/23, Cone NS 14/23
Socket NS 29/32, Cone NS 29/32
Unit Size:
Each
Description
- Connectors with NS stopcock
- With solid glass plug
- With NS socket and NS cone
- Made of DURAN™ tubing
Specifications
Specifications
| Material | Duran Glass |
| Standard Taper Joint No. | Socket NS 29/32, Cone NS 29/32 |