missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Connexin 26 Recombinant Mouse Monoclonal Antibody (24F5)
GREENER_CHOICE

Product Code. 17988912
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17988912 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17988912 Supplier Invitrogen™ Supplier No. MA550243

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Connexin 26 Recombinant Monoclonal Antibody for IHC (P), Flow, ELISA

Gap junctions are conduits that allow the direct cell-to-cell passage of small cytoplasmic molecules, including ions, metabolic intermediates, and second messengers, and thereby mediate intercellular metabolic and electrical communication. Gap junction channels consist of connexin protein subunits, which are encoded by a multigene family. GJBs (gap-junction proteins or connexins) play crucial functional roles associated with these channels. Defects in GJB3 have been linked to erythrokeratodermia variables (EKV) is an autosomal dominant genodermatosis characterized by transient figurate red patches or hyperkeratosis. Mutations in GJB2 have also been associated with genetically derived hearing impairments, including autosomal recessive nonsyndromic deafness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 26
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 24F5
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Gjb2
Gene Accession No. P29033
Gene Alias AI325222; Cnx26; connexin 26; connexin-26; CX26; CXN-26; DFNA3; DFNA3A; DFNB1; DFNB1A; Gap junction beta-2 protein; gap junction channel protein connexin 26; gap junction membrane channel protein beta 2; gap junction protein beta 2; gap junction protein, beta 2; gap junction protein, beta 2, 26kDa; GJB2; Gjb-2; HID; KID; mutant gap junction protein beta 2; NSRD1; PPK
Gene Symbols Gjb2
Host Species Mouse
Immunogen Recombinant Human GJB2 protein.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2706
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.