missing translation for 'onlineSavingsMsg'
Learn More

Connexin 36/GJD2 Antibody - BSA Free, Novus Biologicals™

Product Code. 18676831 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18676831 0.1 mL 0.10mL
18698200 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18676831 Supplier Novus Biologicals Supplier No. NBP2924160.1ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Connexin 36/GJD2 Polyclonal antibody specifically detects Connexin 36/GJD2 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 36/GJD2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Connexin-36, Cx36, CX36connexin 36, Gap junction alpha-9 protein, gap junction delta-2 protein, gap junction protein, alpha 9, 36kDa, gap junction protein, delta 2, 36kDa, GJA9connexin-36, MGC138315, MGC138319
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 99-197 of human GJD2 (NP_065711.1). HQSAKQRERRYSTVFLALDRDPPESIGGPGGTGGGGSGGGKREDKKLQNAIVNGVLQNTENTSKETEPDCLEVKELTPHPSGLRTASKSKLRRQEGISR
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57369
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.