missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Connexin 40.1 Polyclonal specifically detects Connexin 40.1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Connexin 40.1 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | connexin 40.1, Connexin-40.1, Cx40.1, CX40.1connexin40.1, FLJ90023, gap junction delta-4 protein, gap junction protein, delta 4, 40.1kDa, RP11-425A6.2 |
| Gene Symbols | GJD4 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:TDEEGGREEEGAPAPPGARAGGEGAGSPRRTSRVSGHTKIPDEDESEVTSSASEKLGRQPRGRPHREAAQDPRGSG |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?