missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Connexin 40.1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15514772
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15514772 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15514772 Supplier Invitrogen™ Supplier No. PA541321

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: QPRGRPHREA AQDPRGSGSE EQPSAAPSRL AAPPSCSSLQ PPDPPASSSG Sequence homology: Human: 100%.

Connexins, such as GJD4, are involved in the formation of gap junctions, intercellular conduits that directly connect the cytoplasms of contacting cells. Each gap junction channel is formed by docking of 2 hemichannels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 40.1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GJD4
Gene Accession No. Q96KN9
Gene Alias connexin 40.1; connexin40.1; connexin-40.1; CX40.1; Gap junction delta-4 protein; gap junction protein delta 4; gap junction protein, delta 4, 40.1kDa; GJD4
Gene Symbols GJD4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CX40.1 (aa 309-358).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219770
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.