Learn More
Connexin 43 Rabbit, anti-Human, CoraLite Plus 488, Clone: 3O6, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech CL488-80543-20UL
 View more versions of this product
Product Code. 18059855
  • € 140.00 / 20µL
Description
The Connexin 43 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Connexin 43. This antibody recognizes human, mouse, rat, pig antigen. The Connexin 43 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
Connexin 43
3O6
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat, Pig
150 kDa
IgG
Immunofluorescence
1000 μg/mL
Connexin 43, CX43, DFNB38, Gap junction alpha 1 protein, GJA1, GJAL, ODDD
20 μL
2697
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.