missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Connexin 45 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16550612
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16550612 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16550612 Supplier Invitrogen™ Supplier No. PA597784

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Connexin 45 Polyclonal Antibody for ELISA, ICC/IF, Western Blot

Gap junctions were first characterized by electron microscopy as regionally specialized structures on plasma membranes of contacting adherent cells. These structures were shown to consist of cell-to-cell closely packed transmembrane channels. Proteins, called connexins, purified from fractions of enriched gap junctions from different tissues differ. Connexins are designated by their molecular mass. Another system of nomenclature divides gap junction proteins into 2 categories, alpha and beta, according to sequence similarities at the nucleotide and amino acid levels. For example, CX43 is designated alpha-1 gap junction protein, whereas CX32 and CX26 are called beta-1 and beta-2 gap junction proteins, respectively. This nomenclature emphasizes that CX32 and CX26 are more homologous to each other than either of them is to CX43. Connexins have four transmembrane, three intracellular, and two extracellular regions. Different tissues express different connexins, though tissue specificities overlap, and a given tissue or cell can express several different connexins. Developmental regulation of at least some of the connexin genes has been found. Embryo implantation is regulated in part by temporally changing patterns of expression of connexins in the embryo and the maternal decidua.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 45
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene GJC1
Gene Accession No. P36383
Gene Alias C130009G16Rik; Cnx45; connexin 45; connexin-45; CTC-296K1.4; CX45; Cxn-45; Gap junction alpha-7 protein; gap junction channel protein connexin 45; gap junction gamma-1 protein; gap junction membrane channel protein alpha 7; gap junction protein gamma 1; gap junction protein, gamma 1; gap junction protein, gamma 1, 45kDa; Gja7; Gja-7; GJC1
Gene Symbols GJC1
Host Species Rabbit
Immunogen Recombinant Human Gap junction gamma-1 protein (247-396AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10052
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.