Learn More
Emma Safety Footwear Constans Safety Shoes
Click to view available options
Size:
40
46
Unit Size:
1 pair
Description
- Full grain leather upper construction
- Nappa leather lining
- With sole plate for wet work
- Woven zero penetration in-sole
- Antibacterial inlay sole
- Vibram™ Nitril / HRO outsole
Specifications
Specifications
| Color | Black |
| For Use With (Application) | semiorthopedic adjustments requiring customized in-soles |
| Liner Material | Nappa leather |
| Material | Leather |
| Size | 40 |
| Sole Features | Woven zero-penetration insole, Heat-resistant outsole |
| Sole Material | Vibram™ Nitrile |
| Product Type | Constans Safety Shoes |