Learn More
Greiner Bio-One Contact Dishes
Click to view available options
Vent:
No
Yes
Unit Size:
Pack of 600
Description
- Available in different sizes
- With or without vents
- Material: Crystal clear and heat resistant polystyrene
- Contact dish with graduation to enable quick and easy analyses
Specifications
Specifications
| Material | Polystyrene |
| Shape | Round |
| Sterility | Sterile |
| Type | Cell Culture Dish |
| Vent | Yes |
| Height (Metric) | 15 mm |
| Diameter (Metric) | 65 mm |