missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98360AF350
This item is not returnable.
View return policy
Description
Contactin-1 Polyclonal specifically detects Contactin-1 in Mouse samples. It is validated for ELISA.
Specifications
| Contactin-1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1272 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 350 | |
| contactin 1, contactin-1, F3, Glycoprotein gp135, GP135, Neural cell surface protein F3 | |
| Produced in rabbits immunized with purified, recombinant Contactin-1 (NP_031753.1; Met1-Leu1000). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction