missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-3 Antibody (006), Janelia Fluor™ 646 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89312JF646
This item is not returnable.
View return policy
Description
Contactin-3 Monoclonal antibody specifically detects Contactin-3 in Human samples. It is validated for ELISA
Specifications
| Contactin-3 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| contactin 3 (plasmacytoma associated) | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Contactin-3 (Met1-Ser1002,708Asp/Asn; NP_065923.1). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 006 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5067 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction