missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-3 Antibody (524518), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB6644P
This item is not returnable.
View return policy
Description
Contactin-3 Monoclonal antibody specifically detects Contactin-3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| Contactin-3 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5067 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, CyTOF | |
| 524518 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| contactin 3 (plasmacytoma associated) | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Contactin-3, Glu20-Thr1003, Accession # Q07409 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction