missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 415.00
Specifications
| Antigen | Contactin-3 |
|---|---|
| Concentration | 0.05 mg/mL |
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
Description
Contactin-3 Polyclonal specifically detects Contactin-3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Contactin-3 | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| contactin 3 (plasmacytoma associated) | |
| CNTN3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.05 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5067 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ISNLSVTDSGMFQCIAENKHGLVYSSAELKVVASAPDFSKNPMKKLVQVQVGSLVSLDCKPRASPRALSSWKKGDVSVQEHERISLLNDGGLKIANVTKADAGTYTCMAENQFGKANGTTHLVVTEPTRI | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title