missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-5 Antibody (121), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89542B
This item is not returnable.
View return policy
Description
Contactin-5 Monoclonal antibody specifically detects Contactin-5 in Human samples. It is validated for ELISA
Specifications
| Contactin-5 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 121 | |
| ELISA | |
| contactin 5, contactin-5, hNB-2, HNB-2s, MGC163491, NB2, NB-2, neural adhesion molecule, Neural recognition molecule NB-2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Contactin-5 (Met 1-Gln 1059; NP_055176.1). | |
| 0.1 mL | |
| Neuroscience | |
| 53942 | |
| Store at 4°C in the dark. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido