missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-5 Antibody (121), DyLight 550 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89542R
This item is not returnable.
View return policy
Description
Contactin-5 Monoclonal antibody specifically detects Contactin-5 in Human samples. It is validated for ELISA
Specifications
| Contactin-5 | |
| Monoclonal | |
| DyLight 550 | |
| contactin 5, contactin-5, hNB-2, HNB-2s, MGC163491, NB2, NB-2, neural adhesion molecule, Neural recognition molecule NB-2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Contactin-5 (Met 1-Gln 1059; NP_055176.1). | |
| 0.1 mL | |
| Neuroscience | |
| 53942 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 121 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction