missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-5 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97316PCP
This item is not returnable.
View return policy
Description
Contactin-5 Polyclonal antibody specifically detects Contactin-5 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Contactin-5 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| contactin 5, contactin-5, hNB-2, HNB-2s, MGC163491, NB2, NB-2, neural adhesion molecule, Neural recognition molecule NB-2 | |
| Produced in rabbits immunized with E. coli-derived Human Contactin-5 fragment. | |
| 0.1 mL | |
| Neuroscience | |
| 53942 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction