missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Contactin 6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16511872
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16511872 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16511872 Supplier Invitrogen™ Supplier No. PA599047

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Contactin 6 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The neural adhesion molecule Contactin-6, also known as NB-3, is a contactin/F3 subgroup member of immunoglobulin superfamily. It is expressed exclusively in the nervous system and mainly upregulated at the early postnatal stage during mouse brain development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Contactin 6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CNTN6
Gene Accession No. Q9UQ52
Gene Alias CNTN6; contactin 6; contactin-6; hNB-3; mNB-3; NB3; NB-3; neural adhesion molecule; neural recognition molecule NB-3
Gene Symbols CNTN6
Host Species Rabbit
Immunogen Recombinant Human Contactin-6 protein (706-812AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27255
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.