Learn More
Univet™ Contemporary Reading Glasses with Diamond Frame Color
These Contemporary Reading Glasses are lightweight and highly durable while maintaining a glamorous style.
Brand:  Univet™ 571P00598-250
Additional Details : Weight : 0.06800kg
 View more versions of this product
Product Code. 17900872
  • € 150.00 / Pack of 5
Estimated Shipment: 11-06-2024
to see stock.
Description
Description
  • Light and highly resistant
  • Polyamide frame with zero metallic components
  • Integrated side-shields
Specifications
Specifications
Rectangular
None
Diamond
Large
Reusable
Plastic
Standard
Scratch-resistant
Reader Safety Glasses
+2.5
Acrylic
Polycarbonate
UV380