Learn More
SymLine Continuous Level Control Accessories
Click to view available options
Includes:
1 x banana plug, 1 x clamp
1 x clamp, 1 ring ID 10 mm
1 x clamp, 1 x button plug
1 x clamp, 1 x ring ID 5 mm
2 x clamps
2x 10 mm press button
ATEX zone 0, ATEX Ex II 1 G Ex ia IIC T1-T6 Ga
ATEX, II 2 G EEx ia IIC T1-T6, NAMUR
Fitting for continuous level control
Ground cable
Grounding cable and clamp
Product Type:
Antistatic Floor Mat
Capacitive Disc Sensor
Capacitive Rod Sensor
Earthing Cable
Grounding Cable
Grounding Plug
Grounding Strap
Safety Waste Cap
Unit Size:
Each
Beskrivelse
Measuring filling levels continuously - Monitoring non-transparent canisters on a permanent basis - No problem with the continuous level control unit!
Tekniske data
Tekniske data
| Description | Capacitive disc sensor, housing material: stainless steel, Active surface: PTFE, operating distance 1 - 10 mm adjustable, output function: NAMUR. Switch amplifier necessary (108278) |
| Includes | ATEX zone 0, ATEX Ex II 1 G Ex ia IIC T1-T6 Ga |
| Product Type | Capacitive Disc Sensor |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion