Learn More
HUBER Control Cables
Product Code. 10676327
Click to view available options
For Use With (Equipment):
CP AIf / Open
Cc / Rs232 15 Pol
Command Module
Control Cable ECS Standby /Open End
Cp Rs485 / Open
Data Transfer
LEVEL - Open End
PC to Serial Interface
POKO-Alarm/Comgate-Unistat
Tnt&Cc
Unipump/ Unistat
Type:
Control Cable
Control Cable ECS
Control Cable POKO
LEVEL - open end
Unit Size:
Each
Product Code. 10676327
Brand: HUBER 9492
Only null left
Add to basket
Add to basket
Cable Length: 3 m
Cable Length: 3m
Specifications
For Use With (Equipment) LEVEL - Open End
Type LEVEL - open end
Length (Metric) 3 m