Learn More
Eppendorf™ Control Panel
Shop All Eppendorf Products
Click to view available options
:
For use with: For Mastercycler
Description
Reproducibility, specificity and speed - are the requirements for PCR in any application. The Mastercycler pro is unparalleled in its ability to fulfill these requirements. The proprietary vapo. protect™ technology reduces evaporation to a minimum. Concentrations in your PCR master mix remain consistent leading to stable and reproducible specificity. Non-specific binding is minimised beyond importance. The extremely high heating and cooling rates of the Mastercycler pro S give you the speed you need. Unsurpassed speed can be achieved with Eppendorf's Impulse PCR, a device driven hot-start function that increases heating rates to 8°C/s.
- Ultimate reduction of evaporation
- Extremely fast heating and cooling rates
- Gradient blocks with SteadySlope technology
- Intuitive graphic programming
- Display to indicate cycler number in a network
- Upgradable to real-time PCR*
- Optional self-test of peltier elements
Specifications
Specifications
| For Use With (Equipment) | For Mastercycler |
| Product Type | Control Panel |
| Quantity | 1 Ea. |