Learn More
Corning™ CoolBox™ XT Systems
BIOPHARMA
Product Code. 15572791
Click to view available options
Color:
Green
Orange
Pink
Purple
Includes:
CoolBox 2XT System, CoolRack PCR96, SBS-Compatible CoolRack XT M24
CoolBox XT System, SBS-Compatible CoolRack XT M-PCR
CoolBox XT System, SBS-Compatible CoolRack XT M24
Holder and Two Cooling Cores
Holder and one cooling core
Product Line:
CoolBox 2XT
CoolBox 2XT PCR system
CoolBox XT
CoolBox XT M24 system
CoolBox XT PCR Strip system
Unit Size:
Each
Product Code. 15572791
Brand: Corning™ 432022
Only null left
Add to basket
Add to basket
Provide sample cooling or freezing without ice, electricity, or batteries.
Corning CoolBox XT systems are bench top cooling workstations that provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack™ and CoolSink™ thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.
  • Consists of a base, cooling cartridge/core and a lid
  • Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
  • To use with any 30 place Corning CoolRack Tubes Module
  • The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
  • CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
TRUSTED_SUSTAINABILITY
Specifications
Description Benchtop cooling system
Color Green
For Use With (Application) Enables cooling for 16 hours, freezing for 8 hours
Includes Holder and one cooling core
Product Line CoolBox XT