missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolRack™ 96 Storage Tube Modules
BIOPHARMA

Product Code. 15522831
Change view
Click to view available options
Holds:
96 x 0.5 mL 2D Bar Code Tubes
96 x 1.4 mL 2D Bar Code Tubes
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
15522831 96 x 1.4 mL 2D Bar Code Tubes Each
15512831 96 x 0.5 mL 2D Bar Code Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15522831 Supplier Corning™ Supplier No. 432057

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • 2D Barcode storage tube modules
  • Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • Certain CoolRack modules are SBS-compatible.
TRUSTED_SUSTAINABILITY

Specifications

Holds 96 x 1.4 mL 2D Bar Code Tubes
Type Storage Tube Module
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.