missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COP9 signalosome complex subunit 2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-93557CL1
This item is not returnable.
View return policy
Description
COP9 signalosome complex subunit 2 Polyclonal antibody specifically detects COP9 signalosome complex subunit 2 in Drosophila,Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| COP9 signalosome complex subunit 2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Drosophila, Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| ALIEN, Alien homolog, COP9 constitutive photomorphogenic homolog subunit 2 (Arabidopsis), COP9 signalosome complex subunit 2, CSN2JAB1-containing signalosome subunit 2, SGN2TR-interacting protein 15, Signalosome subunit 2, thyroid receptor interacting protein 15, Thyroid receptor-interacting protein 15, TRIP15TRIP-15 | |
| Synthetic peptide of residues 239-250 [ECGGKMHLREGE]. | |
| 0.1 mL | |
| Cell Cycle and Replication, Neuronal Cell Markers, Neurotransmission, Signal Transduction | |
| 9318 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction