missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPB2 Antibody (M3A5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB600-102-0.025ml
This item is not returnable.
View return policy
Description
COPB2 Monoclonal specifically detects COPB2 in Human, Rat, Avian, Bovine, Canine, Chicken, Hamster, Monkey, Primate, Rabbit samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| COPB2 | |
| Monoclonal | |
| Unconjugated | |
| beta prime subunit, Beta'-coat protein, Beta'-COP, betaprime-COP, coatomer protein complex, subunit beta 2 (beta prime), coatomer subunit beta', p102 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Recognizes an epitope shared by the beta-COP protein (110kD) found in most tissue culture cells, and a high MW doublet of brain microtubule-associated protein (MAP 2, 270-300 kD). | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| M3A5 | |
| Western Blot, Immunocytochemistry/Immunofluorescence 1:20-1:50, Immunoprecipitation 1:10-1:500, Immunoaffinity Purification | |
| COPB2 | |
| Microtubule-associated protein from goose brain. | |
| 0.025 mL | |
| Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience | |
| 9276 | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction