missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COPE Polyclonal Antibody

Product Code. 15924635
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15924635 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15924635 Supplier Invitrogen™ Supplier No. PA579059

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat stomach tissue, rat small intestine tissue, rat pancreas tissue, mouse stomach tissue, mouse small intestine tissue, mouse pancreas tissue, human MCF-7 whole cellm, human Hela whole cell, human 22RV1 whole cell. IHC: human mammary cancer tissue, mouse pancreas tissue, rat pancreas tissue. ICC/IF: U20S cell, MCF-7 cell. Flow: HepG2 cell.

The product of this gene is an epsilon subunit of coatomer protein complex. Coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles. It is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. Coatomer complex consists of at least the alpha, beta, beta', gamma, delta, epsilon and zeta subunits. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPE
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene COPE
Gene Accession No. O14579, O89079
Gene Alias 1110005D17Rik; coatomer epsilon subunit; coatomer protein complex subunit epsilon; coatomer protein complex, subunit epsilon; coatomer subunit epsilon; Cope; Cope1; epsilon coat protein; epsilon-coat protein; Epsilon-COP; nonclathrin coat protein epsilon-COP
Gene Symbols COPE
Host Species Rabbit
Immunogen E. coli-derived human COPE recombinant protein (Position: E80-A308).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11316, 290659, 59042
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.