Learn More
Zebrafish Delta D Antibody (zdd2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50372-0.025ml
 View more versions of this product
Product Code. 18674399
  • € 190.00 / 0.03mL
Description
Zebrafish Delta D Monoclonal antibody specifically detects Zebrafish Delta D in Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Zebrafish Delta D
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready
Mouse
Protein A purified
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
zdd2
Unconjugated
PBS
Extracellular domain, Zebrafish Delta D peptide
0.025 mL
Cell Cycle and Replication, Developmental Biology, Neuroscience
Zebrafish